missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-nitrobenzenesulfonyl Chloride 96.0+%, TCI America™
Supplier: TCI America C188725G
Specifications
| 4-Chloro-3-nitrobenzenesulfonyl Chloride | |
| 60°C | |
| C6H3Cl2NO4S | |
| 3261 | |
| SEWNAJIUKSTYOP-UHFFFAOYSA-N | |
| 4-chloro-3-nitrobenzenesulfonyl chloride | |
| 7323 | |
| ≥96.0% (GC,T) |
| 97-08-5 | |
| White-Yellow | |
| 25 g | |
| 4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa | |
| C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl | |
| 256.053 | |
| 256.05 | |
| Crystalline Powder |
Chemical Identifiers
| 97-08-5 | |
| 256.053 | |
| 4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa | |
| 4-chloro-3-nitrobenzenesulfonyl chloride |
| C6H3Cl2NO4S | |
| SEWNAJIUKSTYOP-UHFFFAOYSA-N | |
| 7323 | |
| C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl |
Safety and Handling
EINECSNumber : (3)-2070
TSCA : Yes