Learn More
4-Chloro-3-nitrobenzenesulfonyl chloride, 98%
CAS: 97-08-5 | C6H3Cl2NO4S | 256.053 g/mol
Supplier: Thermo Scientific Chemicals B2163222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chloro-3-nitrobenzenesulfonyl chloride | |
| 58°C to 61°C | |
| MFCD00007440 | |
| UN3261 | |
| Moisture sensitive | |
| SEWNAJIUKSTYOP-UHFFFAOYSA-N | |
| 4-chloro-3-nitrobenzenesulfonyl chloride | |
| 7323 | |
| 98% |
| 97-08-5 | |
| C6H3Cl2NO4S | |
| 100 g | |
| 1978600 | |
| 4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa | |
| C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl | |
| 256.053 | |
| 256.07 |
Chemical Identifiers
| 97-08-5 | |
| 256.053 | |
| SEWNAJIUKSTYOP-UHFFFAOYSA-N | |
| 7323 | |
| C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl |
| C6H3Cl2NO4S | |
| MFCD00007440 | |
| 4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa | |
| 4-chloro-3-nitrobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-558-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only