missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrobenzenesulfonyl chloride, 98%

Catalog No. AAB2163206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2163206 5 g
AAB2163214 25 g
AAB2163222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2163206 Supplier Thermo Scientific Chemicals Supplier No. B2163206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 97-08-5 | C6H3Cl2NO4S | 256.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-08-5
Molecular Formula C6H3Cl2NO4S
Molecular Weight (g/mol) 256.053
MDL Number MFCD00007440
InChI Key SEWNAJIUKSTYOP-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzene-1-sulfonyl chloride, 4-chloro-3-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 4-chloro-3-nitro, yellow sulfon chloride, 3-nitro-4-chlorobenzenesulfonyl chloride, 4-chloro-3-nitrobenzenesulphonyl chloride, 4-chloro-3-nitro benzene sulfonyl chloride, chloro 4-chloro-3-nitrophenyl sulfone, ksc486m2p, sewnajiukstyop-uhfffaoysa
PubChem CID 7323
IUPAC Name 4-chloro-3-nitrobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)[N+](=O)[O-])Cl

Specifications

Melting Point 58°C to 61°C
Quantity 5 g
UN Number UN3261
Beilstein 1978600
Sensitivity Moisture sensitive
Formula Weight 256.07
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-558-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only