missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-iodobenzoic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 42860-04-8 |
|---|---|
| Molecular Formula | C7H4ClIO2 |
| Molecular Weight (g/mol) | 282.46 |
| MDL Number | MFCD00079732 |
| InChI Key | SDRURVZKYHGDAP-UHFFFAOYSA-N |
| Synonym | 3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50 |
| PubChem CID | 2757616 |
| IUPAC Name | 4-chloro-3-iodobenzoic acid |
| SMILES | OC(=O)C1=CC(I)=C(Cl)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC452710050
|
Thermo Scientific Chemicals
452710050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 42860-04-8 | |
| 282.46 | |
| SDRURVZKYHGDAP-UHFFFAOYSA-N | |
| 2757616 | |
| OC(=O)C1=CC(I)=C(Cl)C=C1 |
| C7H4ClIO2 | |
| MFCD00079732 | |
| 3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50 | |
| 4-chloro-3-iodobenzoic acid |
Specifications
| 42860-04-8 | |
| 96% min. (HPLC) | |
| MFCD00079732 | |
| 3-iodo-4-chlorobenzoic acid, 4-chloro-3-iodo-benzoic acid, benzoic acid, 4-chloro-3-iodo, pubchem12500, 4-chloro-3-iodo benzoicacid, benzoic acid,4-chloro-3-iodo, zerenex e/9071913, buttpark 100\01-50 | |
| OC(=O)C1=CC(I)=C(Cl)C=C1 | |
| 282.46 | |
| 282.47 | |
| 4-Chloro-3-iodobenzoic acid |
| Authentic | |
| C7H4ClIO2 | |
| 5g | |
| SDRURVZKYHGDAP-UHFFFAOYSA-N | |
| 4-chloro-3-iodobenzoic acid | |
| 2757616 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO â Research Use Only