missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluoroaniline 97.0+%, TCI America™

Catalog No. C25125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25125G 5 g
C251225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25125G Supplier TCI America Supplier No. C25125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 367-22-6
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD01090987
InChI Key ACMJJQYSPUPMPN-UHFFFAOYSA-N
Synonym 3-fluoro-4-chloroaniline, 4-chloro-3-fluoroanline, 4-chloro-3-fluoro-phenylamine, benzenamine, 4-chloro-3-fluoro, 4-chloro-3-fluorophenylamine, pubchem1516, acmc-1co9a, 4-chloro-3-fluoro-aniline, 4-chloro-3-fluorobenzenamine, ksc226g4r
PubChem CID 2736511
IUPAC Name 4-chloro-3-fluoroaniline
SMILES NC1=CC=C(Cl)C(F)=C1

Specifications

Melting Point 60°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Boiling Point 226°C
Formula Weight 145.56
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 4-Chloro-3-fluoroaniline