missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethoxy)aniline 98.0+%, TCI America™

Catalog No. C3374200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3374200MG 200 mg
C33741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3374200MG Supplier TCI America Supplier No. C3374200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175205-77-3
Molecular Formula C7H5ClF3NO
Molecular Weight (g/mol) 211.568
MDL Number MFCD00153102
InChI Key UCFTYLMKCJPWBX-UHFFFAOYSA-N
PubChem CID 2781526
IUPAC Name 4-chloro-2-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1Cl)OC(F)(F)F)N

Specifications

Melting Point 33°C
Color White-Yellow
Quantity 200 mg
Physical Form Crystal-Lump at 20°C
Synonym 4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine
Boiling Point 82°C
Formula Weight 211.57
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)aniline