missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. C13601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13601G 1 g
C13605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13601G Supplier TCI America Supplier No. C13601G

Chemical Identifiers

CAS 5470-22-4
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00191400
InChI Key NNMYRMGMVLMQAY-UHFFFAOYSA-N
Synonym 4-chloropicolinic acid, 4-chloro-pyridine-2-carboxylic acid, 4-chloro-2-pyridinecarboxylic acid, 4-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 4-chloro, 4-chloro picolinic acid, 2-carboxy-4-chloropyridine, chloropicolinicacid, pubchem12731, acmc-1bn7z
PubChem CID 230890
IUPAC Name 4-chloropyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1Cl)C(=O)O

Specifications

Melting Point 181°C
Color White
Quantity 1 g
Formula Weight 157.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-pyridinecarboxylic Acid