missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7124286
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C021525G 25 g
C0215500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C021525G Supplier TCI America Supplier No. C021525G

Chemical Identifiers

CAS 89-63-4
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00007836
InChI Key PBGKNXWGYQPUJK-UHFFFAOYSA-N
Synonym benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi
PubChem CID 6979
IUPAC Name 4-chloro-2-nitroaniline
SMILES NC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 118°C
Quantity 25 g
UN Number 2237
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-nitroaniline
EINECSNumber (3)-0407&(5)-2303
RTECSNumber BX1575000
TSCA Yes