Learn More
4-chloro-2-nitroaniline, 99%
CAS: 89-63-4 | C6H5ClN2O2 | 172.57 g/mol
$501.87 - $501.87
Chemical Identifiers
| CAS | 89-63-4 |
|---|---|
| Molecular Formula | C6H5ClN2O2 |
| Molecular Weight (g/mol) | 172.57 |
| MDL Number | MFCD00007836 |
| InChI Key | PBGKNXWGYQPUJK-UHFFFAOYSA-N |
| Synonym | benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi |
| PubChem CID | 6979 |
| IUPAC Name | 4-chloro-2-nitroaniline |
| SMILES | NC1=CC=C(Cl)C=C1[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC146520010
|
Thermo Scientific Chemicals
146520010 |
1 kg | Glass bottle |
Each for $501.87
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89-63-4 | |
| 172.57 | |
| PBGKNXWGYQPUJK-UHFFFAOYSA-N | |
| 6979 | |
| NC1=CC=C(Cl)C=C1[N+]([O-])=O |
| C6H5ClN2O2 | |
| MFCD00007836 | |
| benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi | |
| 4-chloro-2-nitroaniline |
Specifications
| 89-63-4 | |
| 100.0 | |
| Orange | |
| Authentic | |
| Glass bottle | |
| ClC6H3(NO2)NH2 | |
| 1 kg | |
| benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi | |
| PBGKNXWGYQPUJK-UHFFFAOYSA-N | |
| 4-chloro-2-nitroaniline | |
| 6979 | |
| 99% | |
| 4-chloro-2-nitroaniline |
| 98.5 | |
| 115°C to 119°C | |
| 191°C | |
| 98.5% min. (GC) | |
| C6H5ClN2O2 | |
| MFCD00007836 | |
| 12, 729 | |
| Solubility in water: insoluble in water. Other solubilities: soluble in methanol and ether | |
| NC1=CC=C(Cl)C=C1[N+]([O-])=O | |
| 172.57 | |
| 172.57 | |
| Crystalline Powder and/or Chunks |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Danger
EINECSNumber : 201-925-4
RUO – Research Use Only