missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-hydroxyacetophenone 96.0+%, TCI America™

Catalog No. p-7125751
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29685G 5 g
C296825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29685G Supplier TCI America Supplier No. C29685G
Only null left

Chemical Identifiers

CAS 6921-66-0
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00238557
InChI Key QCVSDCHNBNFJDQ-UHFFFAOYSA-N
PubChem CID 1051513
IUPAC Name 1-(4-chloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Synonym 1-4-chloro-2-hydroxyphenyl ethanone, 4'-chloro-2'-hydroxyacetophenone, 4-chloro-2-hydroxyacetophenone, 1-4-chloro-2-hydroxyphenyl ethan-1-one, 2'-hydroxy-4'-chloroacetophenone, 1-4-chloro-2-hydroxy-phenyl ethanone, ethanone, 1-4-chloro-2-hydroxyphenyl, 4'-chloro-2'-hydroxy-acetophenone, ethanone,1-4-chloro-2-hydroxyphenyl
Formula Weight 170.59
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Chloro-2′-hydroxyacetophenone