missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-hydroxyacetophenone, 97%

Catalog No. AC449360500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449360500 50 g
AC449360100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449360500 Supplier Thermo Scientific Chemicals Supplier No. 449360500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6921-66-0 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6921-66-0
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
InChI Key QCVSDCHNBNFJDQ-UHFFFAOYSA-N
Synonym 1-4-chloro-2-hydroxyphenyl ethanone, 4'-chloro-2'-hydroxyacetophenone, 4-chloro-2-hydroxyacetophenone, 1-4-chloro-2-hydroxyphenyl ethan-1-one, 2'-hydroxy-4'-chloroacetophenone, 1-4-chloro-2-hydroxy-phenyl ethanone, ethanone, 1-4-chloro-2-hydroxyphenyl, 4'-chloro-2'-hydroxy-acetophenone, ethanone,1-4-chloro-2-hydroxyphenyl
PubChem CID 1051513
IUPAC Name 1-(4-chloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)O

Specifications

Melting Point 27°C to 30°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CLC6H3(OH)COCH3
Quantity 50 g
Solubility Information Solubility in water: insoluble
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2'-hydroxyacetophenone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only