missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-hydroxyacetophenone, 97%

Catalog No. AAH2730909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2730909 10 g
AAH2730918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2730909 Supplier Thermo Scientific Chemicals Supplier No. H2730909

CAS: 6921-66-0 | C8H7ClO2 | 170.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6921-66-0
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00238557
InChI Key QCVSDCHNBNFJDQ-UHFFFAOYSA-N
PubChem CID 1051513
IUPAC Name 1-(4-chloro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)O

Specifications

Melting Point 27°C to 30°C
Quantity 10 g
Synonym 1-4-chloro-2-hydroxyphenyl ethanone, 4'-chloro-2'-hydroxyacetophenone, 4-chloro-2-hydroxyacetophenone, 1-4-chloro-2-hydroxyphenyl ethan-1-one, 2'-hydroxy-4'-chloroacetophenone, 1-4-chloro-2-hydroxy-phenyl ethanone, ethanone, 1-4-chloro-2-hydroxyphenyl, 4'-chloro-2'-hydroxy-acetophenone, ethanone,1-4-chloro-2-hydroxyphenyl
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only