missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluoronitrobenzene, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 455930250
| Quantity | 25g |
|---|
Chemical Identifiers
| C6H3ClFNO2 | |
| MFCD00042211 | |
| 4-chloro-2-fluoronitrobenzene, 2-fluoro-4-chloronitrobenzene, 1-chloro-3-fluoro-4-nitrobenzene, benzene, 4-chloro-2-fluoro-1-nitro, 4-chloro-2-fluoro-1-nitro-benzene, 4-chlor-2-fluor-1-nitrobenzol, pubchem1051, acmc-1bl8e, 4-chloro2-fluoronitrobenzene, ksc380m1b | |
| 4-chloro-2-fluoro-1-nitrobenzene |
Specifications
| 4-Chloro-2-fluoronitrobenzene | |
| >110°C | |
| 97.5% min. (GC) | |
| MFCD00042211 | |
| 4-chloro-2-fluoronitrobenzene, 2-fluoro-4-chloronitrobenzene, 1-chloro-3-fluoro-4-nitrobenzene, benzene, 4-chloro-2-fluoro-1-nitro, 4-chloro-2-fluoro-1-nitro-benzene, 4-chlor-2-fluor-1-nitrobenzol, pubchem1051, acmc-1bl8e, 4-chloro2-fluoronitrobenzene, ksc380m1b | |
| [O-][N+](=O)C1=CC=C(Cl)C=C1F | |
| 175.54 | |
| 175.55 |
| 700-37-8 | |
| Authentic | |
| C6H3ClFNO2 | |
| 25g | |
| PTCPUGKKWNMITF-UHFFFAOYSA-N | |
| 4-chloro-2-fluoro-1-nitrobenzene | |
| 69691 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Call a POISON CENTER or doc
GHS Signal Word: Warning
EINECSNumber : 211-842-5
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.