missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluoroaniline 98.0+%, TCI America™
$92.41 - $180.97
Chemical Identifiers
| CAS | 57946-56-2 |
|---|---|
| Molecular Formula | C6H5ClFN |
| Molecular Weight (g/mol) | 145.56 |
| MDL Number | MFCD00010625 |
| InChI Key | CSFDTBRRIBJILD-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-chloroaniline, 4-chloro-2-fluorobenzenamine, 4-chloro-2-fluoro-phenylamine, benzenamine, 4-chloro-2-fluoro, 4-chloro-2-fluoro-aniline, 4-chloro-2-fluorophenylamine, pubchem1514, 4-chloro-2fluoroaniline, 4chloro-2-fluoroaniline, acmc-1at8a |
| PubChem CID | 93898 |
| IUPAC Name | 4-chloro-2-fluoroaniline |
| SMILES | NC1=CC=C(Cl)C=C1F |
Chemical Identifiers
| 57946-56-2 | |
| 145.56 | |
| CSFDTBRRIBJILD-UHFFFAOYSA-N | |
| 93898 | |
| NC1=CC=C(Cl)C=C1F |
| C6H5ClFN | |
| MFCD00010625 | |
| 2-fluoro-4-chloroaniline, 4-chloro-2-fluorobenzenamine, 4-chloro-2-fluoro-phenylamine, benzenamine, 4-chloro-2-fluoro, 4-chloro-2-fluoro-aniline, 4-chloro-2-fluorophenylamine, pubchem1514, 4-chloro-2fluoroaniline, 4chloro-2-fluoroaniline, acmc-1at8a | |
| 4-chloro-2-fluoroaniline |
Specifications
| 57946-56-2 | |
| 10 g | |
| C6H5ClFN | |
| 2-fluoro-4-chloroaniline, 4-chloro-2-fluorobenzenamine, 4-chloro-2-fluoro-phenylamine, benzenamine, 4-chloro-2-fluoro, 4-chloro-2-fluoro-aniline, 4-chloro-2-fluorophenylamine, pubchem1514, 4-chloro-2fluoroaniline, 4chloro-2-fluoroaniline, acmc-1at8a | |
| NC1=CC=C(Cl)C=C1F | |
| 145.56 | |
| 145.56 | |
| 4-Chloro-2-fluoroaniline |
| 92°C | |
| Clear Liquid at 20°C | |
| MFCD00010625 | |
| CSFDTBRRIBJILD-UHFFFAOYSA-N | |
| 4-chloro-2-fluoroaniline | |
| 93898 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : No