missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoroaniline 98.0+%, TCI America™

Catalog No. C116925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C116925G 25 g
C116910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C116925G Supplier TCI America Supplier No. C116925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57946-56-2
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00010625
InChI Key CSFDTBRRIBJILD-UHFFFAOYSA-N
Synonym 2-fluoro-4-chloroaniline, 4-chloro-2-fluorobenzenamine, 4-chloro-2-fluoro-phenylamine, benzenamine, 4-chloro-2-fluoro, 4-chloro-2-fluoro-aniline, 4-chloro-2-fluorophenylamine, pubchem1514, 4-chloro-2fluoroaniline, 4chloro-2-fluoroaniline, acmc-1at8a
PubChem CID 93898
IUPAC Name 4-chloro-2-fluoroaniline
SMILES NC1=CC=C(Cl)C=C1F

Specifications

Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 92°C
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chloro-2-fluoroaniline