missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoroaniline, 98%

Catalog No. p-7023070
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1613114 25 g
AAA1613122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1613114 Supplier Thermo Scientific Chemicals Supplier No. A1613114
Only null left

CAS: 57946-56-2 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57946-56-2
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00010625
InChI Key CSFDTBRRIBJILD-UHFFFAOYSA-N
Synonym 2-fluoro-4-chloroaniline, 4-chloro-2-fluorobenzenamine, 4-chloro-2-fluoro-phenylamine, benzenamine, 4-chloro-2-fluoro, 4-chloro-2-fluoro-aniline, 4-chloro-2-fluorophenylamine, pubchem1514, 4-chloro-2fluoroaniline, 4chloro-2-fluoroaniline, acmc-1at8a
PubChem CID 93898
IUPAC Name 4-chloro-2-fluoroaniline
SMILES NC1=CC=C(Cl)C=C1F

Specifications

Density 1.311
Quantity 25 g
Refractive Index 1.56
UN Number UN2810
Beilstein 1934804
Boiling Point 92°C (2 mmHg)
Formula Weight 145.56
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 261-034-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only