missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2'-fluoroacetanilide 98.0+%, TCI America™

Catalog No. C16925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16925G 5 g
C169225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16925G Supplier TCI America Supplier No. C16925G
Only null left

Chemical Identifiers

CAS 59280-70-5
Molecular Formula C8H7ClFNO
Molecular Weight (g/mol) 187.598
MDL Number MFCD00042520
InChI Key GVRKNSAEOVXHOS-UHFFFAOYSA-N
Synonym 4'-chloro-2'-fluoroacetanilide, n-4-chloro-2-fluorophenyl acetamide, 4-chloro-2-fluoroacetanilide, 2-fluoro-4-chloroacetanilide, 2'-fluoro-4'-chloroacetanilide, acetamide, n-4-chloro-2-fluorophenyl, acmc-1az4m, 4-chloro2-fluoroacetanilide, 4'-chloro-2'-fluoroacetanilid, 4'-chloro-2'-fluroacetanilide
PubChem CID 853174
IUPAC Name N-(4-chloro-2-fluorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)Cl)F

Specifications

Melting Point 157°C
Quantity 5 g
Formula Weight 187.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloro-2′-fluoroacetanilide