missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-cyanopyridine 98.0+%, TCI America™
$95.06 - $274.02
Chemical Identifiers
| CAS | 19235-89-3 |
|---|---|
| Molecular Formula | C6H3ClN2 |
| Molecular Weight (g/mol) | 138.554 |
| MDL Number | MFCD06009833 |
| InChI Key | DYEZRXLVZMZHQT-UHFFFAOYSA-N |
| Synonym | 4-Chloro-2-pyridinecarbonitrile, 4-Chloropicolinonitrile |
| PubChem CID | 693342 |
| IUPAC Name | 4-chloropyridine-2-carbonitrile |
| SMILES | C1=CN=C(C=C1Cl)C#N |
Chemical Identifiers
| 19235-89-3 | |
| 138.554 | |
| DYEZRXLVZMZHQT-UHFFFAOYSA-N | |
| 693342 | |
| C1=CN=C(C=C1Cl)C#N |
| C6H3ClN2 | |
| MFCD06009833 | |
| 4-Chloro-2-pyridinecarbonitrile, 4-Chloropicolinonitrile | |
| 4-chloropyridine-2-carbonitrile |
Specifications
| 19235-89-3 | |
| C6H3ClN2 | |
| 1 g | |
| 4-Chloro-2-pyridinecarbonitrile, 4-Chloropicolinonitrile | |
| C1=CN=C(C=C1Cl)C#N | |
| 138.554 | |
| 138.55 | |
| Crystalline Powder |
| 84°C | |
| MFCD06009833 | |
| 3439 | |
| DYEZRXLVZMZHQT-UHFFFAOYSA-N | |
| 4-chloropyridine-2-carbonitrile | |
| 693342 | |
| ≥98.0% (GC) | |
| 4-Chloro-2-cyanopyridine |
Safety and Handling
TSCA : No