missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-cyanopyridine 98.0+%, TCI America™

Catalog No. C27325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27325G 5 g
C27321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27325G Supplier TCI America Supplier No. C27325G
Only null left

Chemical Identifiers

CAS 19235-89-3
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.554
MDL Number MFCD06009833
InChI Key DYEZRXLVZMZHQT-UHFFFAOYSA-N
Synonym 4-Chloro-2-pyridinecarbonitrile, 4-Chloropicolinonitrile
PubChem CID 693342
IUPAC Name 4-chloropyridine-2-carbonitrile
SMILES C1=CN=C(C=C1Cl)C#N

Specifications

Melting Point 84°C
Quantity 5 g
UN Number 3439
Formula Weight 138.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-cyanopyridine