missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-cyanopyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 84°C |
| Quantity | 1 g |
| UN Number | 3439 |
| Formula Weight | 138.55 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-2-cyanopyridine |
Safety and Handling
| TSCA | No |