missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2,6-diphenylpyrimidine 98.0+%, TCI America™
$71.23 - $333.05
Chemical Identifiers
| CAS | 29509-91-9 |
|---|---|
| Molecular Formula | C16H11ClN2 |
| Molecular Weight (g/mol) | 266.728 |
| MDL Number | MFCD00234890 |
| InChI Key | MJDDVTZXYXHTRY-UHFFFAOYSA-N |
| PubChem CID | 582041 |
| IUPAC Name | 4-chloro-2,6-diphenylpyrimidine |
| SMILES | C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Cl |
Chemical Identifiers
| 29509-91-9 | |
| 266.728 | |
| MJDDVTZXYXHTRY-UHFFFAOYSA-N | |
| 4-chloro-2,6-diphenylpyrimidine |
| C16H11ClN2 | |
| MFCD00234890 | |
| 582041 | |
| C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Cl |
Specifications
| 29509-91-9 | |
| White | |
| MFCD00234890 | |
| MJDDVTZXYXHTRY-UHFFFAOYSA-N | |
| 4-chloro-2,6-diphenylpyrimidine | |
| 582041 | |
| ≥98.0% (GC) | |
| 4-Chloro-2,6-diphenylpyrimidine |
| 106°C | |
| C16H11ClN2 | |
| 1 g | |
| C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Cl | |
| 266.728 | |
| 266.73 | |
| Crystalline Powder |
Safety and Handling
TSCA : No