missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2,2':6',2″-terpyridine, 99%

Catalog No. AC319670010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319670010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC319670010 Supplier Thermo Scientific Chemicals Supplier No. 319670010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 128143-89-5 | C15H10ClN3 | 267.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128143-89-5
Molecular Formula C15H10ClN3
Molecular Weight (g/mol) 267.72
MDL Number MFCD00191930
InChI Key AHEMFMCEBIJRMU-UHFFFAOYSA-N
Synonym 4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine
PubChem CID 667748
IUPAC Name 4-chloro-2,6-dipyridin-2-ylpyridine
SMILES ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 148.0°C to 152.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 267.72
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Chloro-2, 2':6', 2''-terpyridine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

RUO – Research Use Only