missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2,2':6',2″-terpyridine, 98%

CAS: 128143-89-5 | C15H10ClN3 | 267.72 g/mol

Supplier:  Thermo Scientific Chemicals L1472803

 View more versions of this product

Catalog No. AAL1472803


Only null left
Explore available promotions

Description

Description

4'-Chloro-2,2':6',2″-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture double helicates, dendrimers, micelles and metallo-supramolecular polymers. It is used to synthesize the mono- and bis-terpyridines. It is actively involved in Williamson type ether reactions alpha,μ-bishydroxy-functionalized poly(propylene oxide).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Chloro-2,2′:6′,2′′-terpyridine is widely utilized in the field of supramolecular chemistry, which is used to manufacture double helicates, dendrimers, micelles and metallo-supramolecular polymers. It is used to synthesize the mono- and bis-terpyridines. It is actively involved in Williamson type ether reactions alpha,µ-bishydroxy-functionalized poly(propylene oxide).

Solubility
Slightly soluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

4'-Chloro-2,2':6',2''-terpyridine
148°C to 152°C
MFCD00191930
3613386
Slightly soluble in water.
ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
267.72
267.72
128143-89-5
C15H10ClN3
1 g
4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine
AHEMFMCEBIJRMU-UHFFFAOYSA-N
4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine
667748
98%

Chemical Identifiers

128143-89-5
267.72
AHEMFMCEBIJRMU-UHFFFAOYSA-N
667748
C15H10ClN3
MFCD00191930
4'-chloro-2,2':6',2-terpyridine, 4'-chloro-2,2':6',2'-terpyridine, 4'-chloro-2,2',6',2-terpyridine, 4-chloro-2,6-bis pyridin-2-yl pyridine, 4-chloro-2,6-di 2-pyridyl pyridine, 4-chloro-6-pyridin-2-yl-2,2'-bipyridine, zlchem 975, acmc-1cg8d, 4'-chloro-2,2'6'2-terpyridine, 4-chloro-2,6-bis 2-pyridyl pyridine
ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H301-H315-H318-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only