missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-1H-pyrrolo[2,3-b]pyridine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 179°C |
| Quantity | 200 mg |
| Formula Weight | 152.58 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Chloro-1H-pyrrolo[2,3-b]pyridine |
Safety and Handling
| TSCA | No |