missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1H-pyrazolo[3,4-b]pyridine, 98%

Catalog No. aah66767md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66767MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66767MD Supplier Thermo Scientific Chemicals Supplier No. H66767MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29274-28-0 | C6H4ClN3 | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29274-28-0
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD11846313
InChI Key AKWQSKVILVJWPC-UHFFFAOYSA-N
Synonym 4-chloro-1h-pyrazolo 3,4-b pyridine, 4-chloro-7-aza-1h-indazole, 1h-pyrazolo 3,4-b pyridine, 4-chloro, 1h-pyrazolo 3,4-b pyridine,4-chloro, 5-chloro-2,8,9-triazabicyclo 4.3.0 nona-1,3,5,7-tetraene, 1h-pyrazolo 3, 4-chloro, 4-chloro-7-azabenzpyrazole, 4-chloro-1h-pyrazole 3,4-b pyrimidine, 4-chloranyl-1h-pyrazolo 3,4-b pyridine
PubChem CID 310045
IUPAC Name 4-chloro-1H-pyrazolo[3,4-b]pyridine
SMILES ClC1=C2C=NNC2=NC=C1

Specifications

Quantity 250 mg
Beilstein 607874
Formula Weight 153.57
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-1H-pyrazolo[3,4-b]pyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only