missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-1-methylpiperidine 98.0+%, TCI America™
Supplier: TCI America C246525G
Specifications
| 4-Chloro-1-methylpiperidine | |
| Yellow | |
| C6H12ClN | |
| 25 g | |
| MYGXGCCFTPKWIH-UHFFFAOYSA-N | |
| 4-chloro-1-methylpiperidine | |
| 79342 | |
| ≥98.0% (GC,T) |
| 5570-77-4 | |
| 162°C | |
| MFCD00044489 | |
| 1993 | |
| CN1CCC(Cl)CC1 | |
| 133.62 | |
| 133.62 | |
| Liquid |
Chemical Identifiers
| 5570-77-4 | |
| 133.62 | |
| MYGXGCCFTPKWIH-UHFFFAOYSA-N | |
| 4-chloro-1-methylpiperidine |
| C6H12ClN | |
| MFCD00044489 | |
| 79342 | |
| CN1CCC(Cl)CC1 |
Safety and Handling
TSCA : Yes