missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-1-methylpiperidine Hydrochloride 98.0+%, TCI America™
$42.87 - $128.62
Chemical Identifiers
| CAS | 5382-23-0 |
|---|---|
| Molecular Formula | C6H13Cl2N |
| Molecular Weight (g/mol) | 170.077 |
| MDL Number | MFCD00012834 |
| InChI Key | QDOJNWFOCOWAPO-UHFFFAOYSA-N |
| Synonym | 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n |
| PubChem CID | 3034158 |
| IUPAC Name | 4-chloro-1-methylpiperidine;hydrochloride |
| SMILES | CN1CCC(CC1)Cl.Cl |
Chemical Identifiers
| 5382-23-0 | |
| 170.077 | |
| QDOJNWFOCOWAPO-UHFFFAOYSA-N | |
| 3034158 | |
| CN1CCC(CC1)Cl.Cl |
| C6H13Cl2N | |
| MFCD00012834 | |
| 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n | |
| 4-chloro-1-methylpiperidine;hydrochloride |
Specifications
| 5382-23-0 | |
| White-Yellow | |
| MFCD00012834 | |
| 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n | |
| CN1CCC(CC1)Cl.Cl | |
| 170.077 | |
| 170.08 | |
| Crystalline Powder |
| 162°C | |
| C6H13Cl2N | |
| 25 g | |
| QDOJNWFOCOWAPO-UHFFFAOYSA-N | |
| 4-chloro-1-methylpiperidine;hydrochloride | |
| 3034158 | |
| ≥98.0% (T) | |
| 4-Chloro-1-methylpiperidine Hydrochloride |
Safety and Handling
TSCA : No