missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-methylpiperidine Hydrochloride 98.0+%, TCI America™

Catalog No. C108525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C108525G 25 g
C1085100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C108525G Supplier TCI America Supplier No. C108525G

Chemical Identifiers

CAS 5382-23-0
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012834
InChI Key QDOJNWFOCOWAPO-UHFFFAOYSA-N
Synonym 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
PubChem CID 3034158
IUPAC Name 4-chloro-1-methylpiperidine;hydrochloride
SMILES CN1CCC(CC1)Cl.Cl

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 25 g
Formula Weight 170.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-1-methylpiperidine Hydrochloride