missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-methylpiperidine hydrochloride, 97%

CAS: 5382-23-0 | C6H13Cl2N | 170.077 g/mol

$49.07 - $135.40

Chemical Identifiers

CAS 5382-23-0
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012834
InChI Key QDOJNWFOCOWAPO-UHFFFAOYSA-N
Synonym 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
PubChem CID 3034158
IUPAC Name 4-chloro-1-methylpiperidine;hydrochloride
SMILES CN1CCC(CC1)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0569914
View Documents
Thermo Scientific Chemicals
L0569914
25 g N/A
Only null left
 
AAL0569922
View Documents
Thermo Scientific Chemicals
L0569922
100 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5382-23-0
170.077
QDOJNWFOCOWAPO-UHFFFAOYSA-N
3034158
CN1CCC(CC1)Cl.Cl
C6H13Cl2N
MFCD00012834
4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
4-chloro-1-methylpiperidine;hydrochloride

Specifications

5382-23-0
C6H13Cl2N
25 g
Hygroscopic
QDOJNWFOCOWAPO-UHFFFAOYSA-N
4-chloro-1-methylpiperidine;hydrochloride
3034158
0.97
159°C to 166°C
MFCD00012834
3677071
4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
CN1CCC(CC1)Cl.Cl
170.077
170.08
4-Chloro-1-methylpiperidine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 226-375-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only