missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-methylpiperidine hydrochloride, 97%

Catalog No. AAL0569922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0569922 100 g
AAL0569914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0569922 Supplier Thermo Scientific Chemicals Supplier No. L0569922

CAS: 5382-23-0 | C6H13Cl2N | 170.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5382-23-0
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012834
InChI Key QDOJNWFOCOWAPO-UHFFFAOYSA-N
Synonym 4-chloro-1-methylpiperidine hydrochloride, 1-methyl-4-chloropiperidine hydrochloride, 4-chloro-n-methylpiperidine hydrochloride, 4-chloro-1-methylpiperidinium chloride, piperidine, 4-chloro-1-methyl-, hydrochloride, n-methyl-4-chloropiperidine hydrochloride, 1-methyl-4-chloro piperidine, pubchem2461, acmc-1apdo, ksc495i8n
PubChem CID 3034158
IUPAC Name 4-chloro-1-methylpiperidine;hydrochloride
SMILES CN1CCC(CC1)Cl.Cl

Specifications

Melting Point 159°C to 166°C
Quantity 100 g
Beilstein 3677071
Sensitivity Hygroscopic
Formula Weight 170.08
Percent Purity 0.97
Chemical Name or Material 4-Chloro-1-methylpiperidine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 226-375-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only