missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-butanol, 85%, balance THF and HCl

Catalog No. p-3777864
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC356310100 10 g Glass bottle
AC356311000 100 g Glass bottle
AC356315000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC356310100 Supplier Thermo Scientific Chemicals Supplier No. 356310100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 928-51-8 | C4H9ClO | 108.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-51-8
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00002967
InChI Key HXHGULXINZUGJX-UHFFFAOYSA-N
Synonym 4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
PubChem CID 13569
ChEBI CHEBI:87310
IUPAC Name 4-chlorobutan-1-ol
SMILES OCCCCCl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0890g/mL
Boiling Point 58.0°C to 60.0°C (3.0 mmHg)
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4560
Linear Formula Cl(CH2)4OH
Quantity 10 g
Beilstein 01, II, 398
Specific Gravity 1.089
Formula Weight 108.57
Percent Purity 85%
Physical Form Liquid
Chemical Name or Material 4-Chloro-1-butanol, Balance THF and HCl
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May be corrosive to metals.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Absorb spillage to prevent material damage.
EINECSNumber 213-175-5

RUO – Research Use Only