missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,3-phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America C071025G

 View more versions of this product

Catalog No. C071025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Chloro-1,3-phenylenediamine
5131-60-2
Crystal-Powder at 20°C
MFCD00025284
ZWUBBMDHSZDNTA-UHFFFAOYSA-N
4-chlorobenzene-1,3-diamine
21209
142.59
90°C
25 g
C6H7ClN2
4-chloro-m-phenylenediamine, 1,3-benzenediamine, 4-chloro, 4-chloro-1,3-benzenediamine, 4-chloro-1,3-phenylenediamine, 1-chloro-2,4-diaminobenzene, 4-chloro-meta-phenylenediamine, m-phenylenediamine, 4-chloro, 4-chlorophene-1,3-diamine, 4-chloro-1,3-diaminobenzene, 4-chlorophenylene-1,3-diamine
C1=CC(=C(C=C1N)N)Cl
142.586
CHEBI:82421
≥98.0% (GC,T)

Chemical Identifiers

5131-60-2
142.586
ZWUBBMDHSZDNTA-UHFFFAOYSA-N
21209
4-chlorobenzene-1,3-diamine
C6H7ClN2
MFCD00025284
4-chloro-m-phenylenediamine, 1,3-benzenediamine, 4-chloro, 4-chloro-1,3-benzenediamine, 4-chloro-1,3-phenylenediamine, 1-chloro-2,4-diaminobenzene, 4-chloro-meta-phenylenediamine, m-phenylenediamine, 4-chloro, 4-chlorophene-1,3-diamine, 4-chloro-1,3-diaminobenzene, 4-chlorophenylene-1,3-diamine
CHEBI:82421
C1=CC(=C(C=C1N)N)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0267

RTECSNumber : SS8800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions