missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,3-phenylenediamine 98.0+%, TCI America™

Catalog No. C07105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07105G 5 g
C071025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07105G Supplier TCI America Supplier No. C07105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5131-60-2
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00025284
InChI Key ZWUBBMDHSZDNTA-UHFFFAOYSA-N
Synonym 4-chloro-m-phenylenediamine, 1,3-benzenediamine, 4-chloro, 4-chloro-1,3-benzenediamine, 4-chloro-1,3-phenylenediamine, 1-chloro-2,4-diaminobenzene, 4-chloro-meta-phenylenediamine, m-phenylenediamine, 4-chloro, 4-chlorophene-1,3-diamine, 4-chloro-1,3-diaminobenzene, 4-chlorophenylene-1,3-diamine
PubChem CID 21209
ChEBI CHEBI:82421
IUPAC Name 4-chlorobenzene-1,3-diamine
SMILES C1=CC(=C(C=C1N)N)Cl

Specifications

Melting Point 90°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 142.59
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-Chloro-1,3-phenylenediamine
EINECSNumber (3)-0267
RTECSNumber SS8800000
TSCA Yes