missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,2-phenylenediamine 97.0+%, TCI America™

Catalog No. p-7124314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C02555G 5 g
C025525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C02555G Supplier TCI America Supplier No. C02555G
Only null left

Chemical Identifiers

CAS 95-83-0
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00011691
InChI Key BXIXXXYDDJVHDL-UHFFFAOYSA-N
Synonym 4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine
PubChem CID 7263
ChEBI CHEBI:82301
IUPAC Name 4-chlorobenzene-1,2-diamine
SMILES NC1=CC=C(Cl)C=C1N

Specifications

Melting Point 75°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 142.59
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 4-Chloro-1,2-phenylenediamine
EINECSNumber (3)-0267
RTECSNumber SS8850000
TSCA Yes