missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-1,2-dihydrophthalazin-1-one, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09944DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB09944DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

2257-69-4
180.59
QCKGMJDOJRNSMS-UHFFFAOYSA-N
2775142
ClC1=NNC(=O)C2=CC=CC=C12
C8H5ClN2O
MFCD00102192
4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one
4-chloro-2H-phthalazin-1-one

Specifications

4-chloro-1, 2-dihydrophthalazin-1-one
96.5
97%
C8H5ClN2O
MFCD00102192
QCKGMJDOJRNSMS-UHFFFAOYSA-N
4-chloro-2H-phthalazin-1-one
2775142
97%
2257-69-4
100.0
Amber Glass Bottle
5g
4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one
ClC1=NNC(=O)C2=CC=CC=C12
180.59
180.59
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents