missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$53.90 - $53.90
Chemical Identifiers
| CAS | 14047-29-1 |
|---|---|
| Molecular Formula | C7H7BO4 |
| MDL Number | MFCD00151801 |
| InChI Key | SIAVMDKGVRXFAX-UHFFFAOYSA-N |
| PubChem CID | 312183 |
Specifications
| 14047-29-1 | |
| C7H7BO4 | |
| 1 g | |
| SIAVMDKGVRXFAX-UHFFFAOYSA-N | |
| 4-(dihydroxyboranyl)benzoic acid | |
| 312183 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00151801 | |
| 4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324 | |
| OB(O)C1=CC=C(C=C1)C(O)=O | |
| 165.94 | |
| 165.94 | |
| 4-Carboxyphenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
RTECSNumber : CY8925000
TSCA : No