missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C13531G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13531G Supplier TCI America Supplier No. C13531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14047-29-1
Molecular Formula C7H7BO4
MDL Number MFCD00151801
InChI Key SIAVMDKGVRXFAX-UHFFFAOYSA-N
PubChem CID 312183

Specifications

Color White-Yellow
Quantity 1 g
Synonym 4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
SMILES OB(O)C1=CC=C(C=C1)C(O)=O
IUPAC Name 4-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol) 165.94
Formula Weight 165.94
Physical Form Crystalline Powder
Chemical Name or Material 4-Carboxyphenylboronic Acid (contains varying amounts of Anhydride)
RTECSNumber CY8925000
TSCA No