missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine hydrochloride, 97%, anhydrous

Catalog No. AC434850010
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434850010 1 kg Glass bottle
AC434850250 25 g Glass bottle
AC434851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC434850010 Supplier Thermo Scientific Chemicals Supplier No. 434850010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52-89-1 | C3H8ClNO2S | 157.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52-89-1
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064553
InChI Key IFQSXNOEEPCSLW-QYLAHFPANA-N
Synonym l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
PubChem CID 60960
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES Cl.N[C@@H](CS)C(O)=O

Specifications

Melting Point 175.0°C to 181.0°C
Color White to Brown
Elemental Analysis 22.29 to 23.43 % C 8.67 to 9.11 % N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HSCH2CH(NH2)CO2H·HCl
Quantity 1 kg
Merck Index 15,2778
Specific Rotation + 7.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Formula Weight 157.61
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Cysteine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-157-7

RUO – Research Use Only