missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzeneboronic acid, 97%

CAS: 14047-29-1 | C7H7BO4 | 165.94 g/mol

Supplier:  Thermo Scientific Chemicals B2095406

 View more versions of this product

Catalog No. AAB2095406

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Carboxybenzeneboronic acid
∼220°C (decomposition)
MFCD00151801
3031088
SIAVMDKGVRXFAX-UHFFFAOYSA-N
4-boronobenzoic acid
312183
97%
14047-29-1
C7H7BO4
5 g
4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
OB(O)C1=CC=C(C=C1)C(O)=O
165.94
165.94

Chemical Identifiers

14047-29-1
165.94
SIAVMDKGVRXFAX-UHFFFAOYSA-N
312183
OB(O)C1=CC=C(C=C1)C(O)=O
C7H7BO4
MFCD00151801
4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
4-boronobenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

RTECSNumber : CY8925000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only