missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzeneboronic acid, 97%

Catalog No. AAB2095414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2095414 25 g
AAB2095403 1 g
AAB2095406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2095414 Supplier Thermo Scientific Chemicals Supplier No. B2095414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14047-29-1 | C7H7BO4 | 165.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14047-29-1
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.94
MDL Number MFCD00151801
InChI Key SIAVMDKGVRXFAX-UHFFFAOYSA-N
Synonym 4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
PubChem CID 312183
IUPAC Name 4-boronobenzoic acid
SMILES OB(O)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point ∼220°C (decomposition)
Quantity 25 g
Beilstein 3031088
Formula Weight 165.94
Percent Purity 97%
Chemical Name or Material 4-Carboxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber CY8925000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only