missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzaldehyde, 96%

Catalog No. p-3735820
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154580050 5 g Glass bottle
AC154580250 25 g Glass bottle
AC154581000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154580050 Supplier Thermo Scientific Chemicals Supplier No. 154580050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 619-66-9 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-66-9
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00006951
InChI Key GOUHYARYYWKXHS-UHFFFAOYSA-N
Synonym 4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
PubChem CID 12088
IUPAC Name 4-formylbenzoic acid
SMILES C1=CC(=CC=C1C=O)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 247°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula HO2CC6H4CHO
Quantity 5 g
Beilstein 10, 671
Solubility Information Solubility in water: soluble
Formula Weight 150.13
Percent Purity 96%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Carboxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-607-4

RUO – Research Use Only