missing translation for 'onlineSavingsMsg'
Learn More

4-Butyrylbiphenyl 99.0+%, TCI America™

Catalog No. p-7119380
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
B195625G 25 g
B1956500G 500 g
2 options

Catalog No. B195625G

Supplier: TCI America B195625G

Only null left

Chemical Identifiers

CAS 13211-01-3
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
MDL Number MFCD00022005
InChI Key WOFNMZNJGZJHJO-UHFFFAOYSA-N
PubChem CID 83235
IUPAC Name 1-(4-phenylphenyl)butan-1-one
SMILES CCCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 95°C
Boiling Point 355°C
Quantity 25 g
Synonym 4′C-Phenylbutyrophenone
Formula Weight 224.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butyrylbiphenyl