missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
$333.67 - $984.22
Chemical Identifiers
| CAS | 71101-89-8 |
|---|---|
| Molecular Formula | C11H20O2 |
| Molecular Weight (g/mol) | 184.279 |
| MDL Number | MFCD00460757 |
| InChI Key | BALGERHMIXFENA-UHFFFAOYSA-N |
| PubChem CID | 2060518 |
| IUPAC Name | 4-butylcyclohexane-1-carboxylic acid |
| SMILES | CCCCC1CCC(CC1)C(=O)O |
Chemical Identifiers
| 71101-89-8 | |
| 184.279 | |
| BALGERHMIXFENA-UHFFFAOYSA-N | |
| 4-butylcyclohexane-1-carboxylic acid |
| C11H20O2 | |
| MFCD00460757 | |
| 2060518 | |
| CCCCC1CCC(CC1)C(=O)O |
Specifications
| 71101-89-8 | |
| C11H20O2 | |
| 5 g | |
| CCCCC1CCC(CC1)C(=O)O | |
| 184.279 | |
| 184.28 | |
| Liquid |
| Yellow | |
| MFCD00460757 | |
| BALGERHMIXFENA-UHFFFAOYSA-N | |
| 4-butylcyclohexane-1-carboxylic acid | |
| 2060518 | |
| ≥98.0% (GC,T) | |
| 4-Butylcyclohexanecarboxylic Acid (cis- and trans- mixture) |
Safety and Handling
EINECSNumber : (3)-3723
TSCA : No