missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butylbenzoyl Chloride 96.0+%, TCI America™
$413.83 - $413.83
Chemical Identifiers
| CAS | 28788-62-7 |
|---|---|
| Molecular Formula | C11H13ClO |
| Molecular Weight (g/mol) | 196.67 |
| MDL Number | MFCD00000699 |
| InChI Key | OUOWCSJYDCPVDM-UHFFFAOYSA-N |
| Synonym | 4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o |
| PubChem CID | 120049 |
| IUPAC Name | 4-butylbenzoyl chloride |
| SMILES | CCCCC1=CC=C(C=C1)C(Cl)=O |
Chemical Identifiers
| 28788-62-7 | |
| 196.67 | |
| OUOWCSJYDCPVDM-UHFFFAOYSA-N | |
| 120049 | |
| CCCCC1=CC=C(C=C1)C(Cl)=O |
| C11H13ClO | |
| MFCD00000699 | |
| 4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o | |
| 4-butylbenzoyl chloride |
Specifications
| 28788-62-7 | |
| C11H13ClO | |
| 25 g | |
| 4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o | |
| CCCCC1=CC=C(C=C1)C(Cl)=O | |
| 196.67 | |
| 196.67 | |
| Liquid |
| Yellow | |
| MFCD00000699 | |
| 3265 | |
| OUOWCSJYDCPVDM-UHFFFAOYSA-N | |
| 4-butylbenzoyl chloride | |
| 120049 | |
| ≥96.0% (GC,T) | |
| 4-Butylbenzoyl Chloride |
Safety and Handling
EINECSNumber : (3)-3545
TSCA : Yes