missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Butylacetophenone 97.0+%, TCI America™
$167.49 - $167.49
Chemical Identifiers
| CAS | 37920-25-5 |
|---|---|
| Molecular Formula | C12H16O |
| Molecular Weight (g/mol) | 176.26 |
| MDL Number | MFCD00017500 |
| InChI Key | MQESVSITPLILCO-UHFFFAOYSA-N |
| Synonym | 4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone |
| PubChem CID | 123470 |
| IUPAC Name | 1-(4-butylphenyl)ethan-1-one |
| SMILES | CCCCC1=CC=C(C=C1)C(C)=O |
Chemical Identifiers
| 37920-25-5 | |
| 176.26 | |
| MQESVSITPLILCO-UHFFFAOYSA-N | |
| 123470 | |
| CCCCC1=CC=C(C=C1)C(C)=O |
| C12H16O | |
| MFCD00017500 | |
| 4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone | |
| 1-(4-butylphenyl)ethan-1-one |
Specifications
| 37920-25-5 | |
| 270°C | |
| MFCD00017500 | |
| 4'-butylacetophenone, 1-4-butylphenyl ethanone, ethanone, 1-4-butylphenyl, p-butylacetophenone, 1-4-butylphenyl ethan-1-one, p-n-butylacetophenone, 4'-n-butylacetophenone, unii-6738cf9z8v, 1-acetyl-4-butylbenzene, 1-4-butyl-phenyl ethanone | |
| CCCCC1=CC=C(C=C1)C(C)=O | |
| 176.26 | |
| 176.26 | |
| Liquid |
| Yellow | |
| C12H16O | |
| 25 g | |
| MQESVSITPLILCO-UHFFFAOYSA-N | |
| 1-(4-butylphenyl)ethan-1-one | |
| 123470 | |
| ≥97.0% (GC) | |
| 4′-Butylacetophenone |
Safety and Handling
EINECSNumber : (3)-3540
TSCA : Yes