missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$63.38 - $63.38
Chemical Identifiers
| CAS | 105365-51-3 |
|---|---|
| Molecular Formula | C10H15BO3 |
| MDL Number | MFCD03427054 |
| InChI Key | QUPFQMXWFNJUNJ-UHFFFAOYSA-N |
| PubChem CID | 3836310 |
| IUPAC Name | (4-butoxyphenyl)boronic acid |
Chemical Identifiers
| 105365-51-3 | |
| MFCD03427054 | |
| 3836310 |
| C10H15BO3 | |
| QUPFQMXWFNJUNJ-UHFFFAOYSA-N | |
| (4-butoxyphenyl)boronic acid |
Specifications
| 105365-51-3 | |
| White | |
| MFCD03427054 | |
| 4-n-butoxyphenylboronic acid, 4-butoxyphenyl boronic acid, 4-n-butoxyphenyl boronic acid, p-butoxyphenylboronic acid, 4-butyloxyphenylboronic acid, 4-n-butoxy benzeneboronic acid, 4-butoxymethylboronic acid, boronic acid, 4-butoxyphenyl, pubchem9549, 4-butoxybenzeneboronic acid | |
| CCCCOC1=CC=C(C=C1)B(O)O | |
| 194.04 | |
| 194.04 | |
| 4-Butoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 108°C | |
| C10H15BO3 | |
| 1 g | |
| QUPFQMXWFNJUNJ-UHFFFAOYSA-N | |
| (4-butoxyphenyl)boronic acid | |
| 3836310 | |
| Crystalline Powder |
Safety and Handling
TSCA : No