missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B47271G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47271G Supplier TCI America Supplier No. B47271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105365-51-3
Molecular Formula C10H15BO3
MDL Number MFCD03427054
InChI Key QUPFQMXWFNJUNJ-UHFFFAOYSA-N
PubChem CID 3836310
IUPAC Name (4-butoxyphenyl)boronic acid

Specifications

Melting Point 108°C
Color White
Quantity 1 g
Synonym 4-n-butoxyphenylboronic acid, 4-butoxyphenyl boronic acid, 4-n-butoxyphenyl boronic acid, p-butoxyphenylboronic acid, 4-butyloxyphenylboronic acid, 4-n-butoxy benzeneboronic acid, 4-butoxymethylboronic acid, boronic acid, 4-butoxyphenyl, pubchem9549, 4-butoxybenzeneboronic acid
SMILES CCCCOC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 194.04
Formula Weight 194.04
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxyphenylboronic Acid (contains varying amounts of Anhydride)