missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromovalerophenone 98.0+%, TCI America™

Catalog No. B47335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47335G 5 g
B473325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47335G Supplier TCI America Supplier No. B47335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7295-44-5
Molecular Formula C11H13BrO
Molecular Weight (g/mol) 241.128
MDL Number MFCD00000107
InChI Key STYJKBMRWQQJIS-UHFFFAOYSA-N
Synonym 4'-bromovalerophenone, 1-4-bromophenyl pentan-1-one, p-bromophenyl butyl ketone, p-bromovalerophenone, 1-pentanone, 1-4-bromophenyl, 4-bromovalerophenone, 1-bromo-4-pentanoylbenzene, 1-4-bromophenyl-1-pentanone, acmc-20ammf, 4;-bromovalerophenone
PubChem CID 81717
IUPAC Name 1-(4-bromophenyl)pentan-1-one
SMILES CCCCC(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 170°C
Quantity 5 g
Formula Weight 241.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromovalerophenone
TSCA No
Recommended Storage Refrigerator