missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromotriphenylamine 97.0+%, TCI America™
$63.52 - $180.97
Chemical Identifiers
| CAS | 36809-26-4 |
|---|---|
| Molecular Formula | C18H14BrN |
| Molecular Weight (g/mol) | 324.221 |
| InChI Key | SQTLUXJWUCHKMT-UHFFFAOYSA-N |
| Synonym | 4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene |
| PubChem CID | 642838 |
| IUPAC Name | 4-bromo-N,N-diphenylaniline |
| SMILES | C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br |
Chemical Identifiers
| 36809-26-4 | |
| 324.221 | |
| 4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene | |
| 4-bromo-N,N-diphenylaniline |
| C18H14BrN | |
| SQTLUXJWUCHKMT-UHFFFAOYSA-N | |
| 642838 | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br |
Specifications
| 36809-26-4 | |
| White-Yellow | |
| C18H14BrN | |
| 4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br | |
| 324.221 | |
| 324.22 | |
| Crystalline Powder |
| 115°C | |
| 185°C | |
| 5 g | |
| SQTLUXJWUCHKMT-UHFFFAOYSA-N | |
| 4-bromo-N,N-diphenylaniline | |
| 642838 | |
| ≥97.0% (GC) | |
| 4-Bromotriphenylamine |
Safety and Handling
TSCA : No