missing translation for 'onlineSavingsMsg'
Learn More

4-Bromotriphenylamine, 99%

Catalog No. AAH6032806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6032806 5 g
AAH6032814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6032806 Supplier Thermo Scientific Chemicals Supplier No. H6032806

CAS: 36809-26-4 | C18H14BrN | 324.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36809-26-4
Molecular Formula C18H14BrN
Molecular Weight (g/mol) 324.221
MDL Number MFCD01851266
InChI Key SQTLUXJWUCHKMT-UHFFFAOYSA-N
Synonym 4-bromotriphenylamine, 4-bromo-phenyl-diphenyl-amine, 4-bromophenyl diphenylamine, 4-bromo triphenylamine, benzenamine, 4-bromo-n,n-diphenyl, n,n-diphenyl-4-bromoaniline, rarechem fh 1g 0a02, n-4-bromophenyl-n-phenylbenzenamine, 4-diphenylamino-1-bromobenzene
PubChem CID 642838
IUPAC Name 4-bromo-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 108°C to 112°C
Quantity 5 g
Formula Weight 324.21
Percent Purity 99%
Chemical Name or Material 4-Bromotriphenylamine
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only